2,3,4-Trifluorophenacyl bromide

Lieferant: Apollo Scientific
Gefahr

PC49998-1G PC49998-250MG PC49998-5G
APOSPC49998-1GEA 522 EUR
APOSPC49998-1G APOSPC49998-250MG APOSPC49998-5G
2,3,4-Trifluorophenacyl bromide
2,3,4-Trifluorophenacyl bromide
Formel: C₈H₄BrF₃O
Molecular Weight: 253.016 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194476
CAS-Nummer: 1214345-02-4
UN: 1759
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR