5-Bromo-3,4-difluorobenzene-1,2-diamine
Lieferant: Apollo Scientific
PC500007-1G
PC500007-250MG
APOSPC500007-1GEA
706
EUR
APOSPC500007-1G
APOSPC500007-250MG
5-Bromo-3,4-difluorobenzene-1,2-diamine
5-Bromo-3,4-difluorobenzene-1,2-diamine
Formel:
C₆H₅BrF₂N₂ Molecular Weight: 223,018 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22375091 CAS-Nummer: 1210048-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...