6-(Trifluoromethyl)isatoic anhydride 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Trifluoromethyl)-2H-3,1-benzoxazine-2,4(1H)-dione, 5-(Trifluoromethyl)-1H-benzo[d][1,3]oxazine-2,4-dione
PC500009-1G
PC500009-250MG
APOSPC500009-1GEA
322
EUR
APOSPC500009-1G
APOSPC500009-250MG
6-(Trifluoromethyl)isatoic anhydride 97%
6-(Trifluoromethyl)isatoic anhydride
Formel:
C₉H₄F₃NO₃ Molecular Weight: 231,128 g/mol Schmelzpunkt: 239 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD24850066 CAS-Nummer: 387-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...