5-(Trifluoromethyl)isatoic anhydride

Lieferant: Apollo Scientific
Achtung

PC500012-1G PC500012-250MG PC500012-5G PC500012-25G
APOSPC500012-1GEA 0 EUR
APOSPC500012-1G APOSPC500012-250MG APOSPC500012-5G APOSPC500012-25G
5-(Trifluoromethyl)isatoic anhydride
5-(Trifluoromethyl)isatoic anhydride
Formel: C₉H₄F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD20501921
CAS-Nummer: 781-94-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR