4,5-Difluoroisatoic anhydride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6,7-Difluoro-1H-benzo[d][1,3]oxazine-2,4-dione, 6,7-Difluoro-2H-3,1-benzoxazine-2,4(1H)-dione
PC500013-1G
PC500013-250MG
APOSPC500013-1GEA
301
EUR
APOSPC500013-1G
APOSPC500013-250MG
4,5-Difluoroisatoic anhydride
4,5-Difluoroisatoic anhydride
Formel:
C₈H₃F₂NO₃ Molecular Weight: 199,111 g/mol Schmelzpunkt: 223…226 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08457975 CAS-Nummer: 97927-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...