5-Bromo-4-fluoroisatoic anhydride 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-7-fluoro-2H-3,1-benzoxazine-2,4(1H)-dione, 6-Bromo-7-fluoro-1H-benzo[d][1,3]oxazine-2,4-dione
PC500014-1G
PC500014-250MG
PC500014-5G
PC500014-25G
PC500014-100G
APOSPC500014-1GEA
119
EUR
APOSPC500014-1G
APOSPC500014-250MG
APOSPC500014-5G
APOSPC500014-25G
APOSPC500014-100G
5-Bromo-4-fluoroisatoic anhydride 95%
5-Bromo-4-fluoroisatoic anhydride
Formel:
C₈H₃BrFNO₃ Molecular Weight: 260,017 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24850064 CAS-Nummer: 1440535-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...