3-(Trifluoromethyl)isatoic anhydride
Lieferant: Apollo Scientific
|
Achtung
|
PC500017-1G
PC500017-250MG
APOSPC500017-1GEA
847
EUR
APOSPC500017-1G
APOSPC500017-250MG
3-(Trifluoromethyl)isatoic anhydride
3-(Trifluoromethyl)isatoic anhydride
Formel:
C₉H₄F₃NO₃ Molecular Weight: 231,128 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18251388 CAS-Nummer: 72985-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...