4-Nitro-3-(trifluoromethyl)phenyl trifluoromethanesulphonate
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Nitro-3-(trifluoromethyl)phenyl triflate, 2-Nitro-5-{[(trifluoromethyl)sulphonyl]oxy}benzotrifluoride
PC500115-1G
PC500115-250MG
APOSPC500115-1GEA
154
EUR
APOSPC500115-1G
APOSPC500115-250MG
4-Nitro-3-(trifluoromethyl)phenyl trifluoromethanesulphonate
4-Nitro-3-(trifluoromethyl)phenyl trifluoromethanesulphonate
Formel:
C₈H₃F₆NO₅S Molecular Weight: 339,169 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25541875 CAS-Nummer: 1446016-89-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...