2-Fluoro-3-methoxyphenol 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Fluoro-3-hydroxyanisole
PC500160-5G
PC500160-100G
PC500160-25G
APOSPC500160-5GEA
132
EUR
APOSPC500160-5G
APOSPC500160-100G
APOSPC500160-25G
2-Fluoro-3-methoxyphenol 98%
2-Fluoro-3-methoxyphenol
Formel:
C₇H₇FO₂ Molecular Weight: 142,128 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11110186 CAS-Nummer: 447462-87-5 UN: 1759 ADR: 8,III |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...