4-Fluoro-3-methoxyphenol 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-5-hydroxyanisole

PC500164-1G PC500164-5G PC500164-25G
APOSPC500164-1GEA 23.2 EUR
APOSPC500164-1G APOSPC500164-5G APOSPC500164-25G
4-Fluoro-3-methoxyphenol 96%
4-Fluoro-3-methoxyphenol
Formel: C₇H₇FO₂
Molecular Weight: 142,128 g/mol
Siedepunkt: 125 °C (0,75 mm Hg)
Lagertemperatur: Kühlschrank
MDL: MFCD10698649
CAS-Nummer: 117902-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR