5-Bromo-4-fluoro-2-nitroanisole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-4-fluoro-2-nitrophenyl methyl ether, 1-Bromo-2-fluoro-5-methoxy-4-nitrobenzene, 4-Bromo-5-fluoro-2-methoxynitrobenzene
PC500181-1G
PC500181-25G
PC500181-5G
APOSPC500181-1GEA
36.4
EUR
APOSPC500181-1G
APOSPC500181-25G
APOSPC500181-5G
5-Bromo-4-fluoro-2-nitroanisole
5-Bromo-4-fluoro-2-nitroanisole
Formel:
C₇H₅BrFNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD16658620 CAS-Nummer: 330794-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...