2-Amino-4-bromo-3-fluorobenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-6-carboxy-2-fluoroaniline, 2-amino-4-bromo-3-fluorobenzoic acid, 4-Bromo-3-fluoroanthranilic acid

PC500189-5G PC500189-1G PC500189-250MG
APOSPC500189-5GEA 418 EUR
APOSPC500189-5G APOSPC500189-1G APOSPC500189-250MG
2-Amino-4-bromo-3-fluorobenzoic acid 95%
2-Amino-4-bromo-3-fluorobenzoic acid
Formel: C₇H₅BrFNO₂
Molecular Weight: 234.023 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD27664889
CAS-Nummer: 1416013-62-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR