2',4'-Difluoro-5'-(trifluoromethyl)acetophenone 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-[2,4-Difluoro-5-(trifluoromethyl)phenyl]ethanone
PC500191-1G
PC500191-250MG
APOSPC500191-1GEA
69.6
EUR
APOSPC500191-1G
APOSPC500191-250MG
2',4'-Difluoro-5'-(trifluoromethyl)acetophenone 95%
2',4'-Difluoro-5'-(trifluoromethyl)acetophenone
Formel:
C₉H₅F₅O Lagertemperatur: Raumtemperatur |
MDL:
MFCD27664904 CAS-Nummer: 1288987-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...