3-Bromo-2-fluoro-6-nitrophenylacetic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-(carboxymethyl)-3-fluoronitrobenzene, (3-Bromo-2-fluoro-6-nitrophenyl)acetic acid

PC500196-1G PC500196-250MG
APOSPC500196-1GEA 905 EUR
APOSPC500196-1G APOSPC500196-250MG
3-Bromo-2-fluoro-6-nitrophenylacetic acid 95%
3-Bromo-2-fluoro-6-nitrophenylacetic acid
Formel: C₈H₅BrFNO₄
Molecular Weight: 278.032 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27664856
CAS-Nummer: 1807155-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR