2,6-Difluoro-3-(trifluoromethyl)benzonitrile

Lieferant: Apollo Scientific
Gefahr

PC500219-1G PC500219-250MG PC500219-5G
APOSPC500219-1GEA 215 EUR
APOSPC500219-1G APOSPC500219-250MG APOSPC500219-5G
2,6-Difluoro-3-(trifluoromethyl)benzonitrile
2,6-Difluoro-3-(trifluoromethyl)benzonitrile
Formel: C₈H₂F₅N
Molecular Weight: 207,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27937181
CAS-Nummer: 1806333-92-5
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR