2,6-Difluoro-3-(trifluoromethyl)phenacyl bromide

Lieferant: Apollo Scientific
Gefahr

PC500240-1G PC500240-250MG
APOSPC500240-1GEA 654 EUR
APOSPC500240-1G APOSPC500240-250MG
2,6-Difluoro-3-(trifluoromethyl)phenacyl bromide
2,6-Difluoro-3-(trifluoromethyl)phenacyl bromide
Formel: C₉H₄BrF₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD27937167
CAS-Nummer: 1823271-81-3
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR