2-Bromo-4-fluoro-5-nitroanisole 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-5-fluoro-2-methoxy-4-nitrobenzene
PC500265-1G
PC500265-250MG
PC500265-5G
APOSPC500265-1GEA
456
EUR
APOSPC500265-1G
APOSPC500265-250MG
APOSPC500265-5G
2-Bromo-4-fluoro-5-nitroanisole 95%
2-Bromo-4-fluoro-5-nitroanisole
Formel:
C₇H₅BrFNO₃ Molecular Weight: 250.022 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27975461 CAS-Nummer: 84478-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...