6-Fluoro-2-hydroxy-3-nitrobenzaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Fluoro-2-formyl-6-nitrophenol, 4-Fluoro-3-formyl-2-hydroxynitrobenzene
PC500274-1G
PC500274-250MG
APOSPC500274-1GEA
383
EUR
APOSPC500274-1G
APOSPC500274-250MG
6-Fluoro-2-hydroxy-3-nitrobenzaldehyde
6-Fluoro-2-hydroxy-3-nitrobenzaldehyde
Formel:
C₇H₄FNO₄ Molecular Weight: 185,109 g/mol Schmelzpunkt: 80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD27975498 CAS-Nummer: 38225-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...