Methyl 2,3-diamino-6-fluorobenzoate

Lieferant: Apollo Scientific
Achtung

PC500283-1G PC500283-250MG
APOSPC500283-1GEA 476 EUR
APOSPC500283-1G APOSPC500283-250MG
Methyl 2,3-diamino-6-fluorobenzoate
Methyl 2,3-diamino-6-fluorobenzoate
Formel: C₈H₉FN₂O₂
Molecular Weight: 184,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20701664
CAS-Nummer: 403712-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR