7-Bromo-5-fluoro-1,3-benzoxazole 95%

Lieferant: Apollo Scientific
Achtung

PC500297-1G PC500297-250MG PC500297-5G
APOSPC500297-1GEA 493 EUR
APOSPC500297-1G APOSPC500297-250MG APOSPC500297-5G
7-Bromo-5-fluoro-1,3-benzoxazole 95%
7-Bromo-5-fluoro-1,3-benzoxazole
Formel: C₇H₃BrFNO
Molecular Weight: 216,007 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16468547
CAS-Nummer: 1226045-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR