5-Methyl-2-(trifluoromethyl)-3-furoyl chloride 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Methyl-2-(trifluoromethyl)furan-3-carbonyl chloride, 3-(Chlorocarbonyl)-5-methyl-2-(trifluoromethyl)furan
PC5003-1G
PC5003-2.5G
APOSPC5003-1GEA
200
EUR
APOSPC5003-1G
APOSPC5003-2.5G
5-Methyl-2-(trifluoromethyl)-3-furoyl chloride 95+%
5-Methyl-2-(trifluoromethyl)-3-furoyl chloride
Formel:
C₇H₄ClF₃O₂ Molecular Weight: 212.554 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00275562 CAS-Nummer: 175276-66-1 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...