2-Fluoro-4-methyl-5-nitrophenol
Lieferant: Apollo Scientific
|
Achtung
|
PC500302-1G
PC500302-250MG
APOSPC500302-1GEA
177
EUR
APOSPC500302-1G
APOSPC500302-250MG
2-Fluoro-4-methyl-5-nitrophenol
2-Fluoro-4-methyl-5-nitrophenol
Formel:
C₇H₆FNO₃ Molecular Weight: 171.126 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22586680 CAS-Nummer: 110298-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...