2-Fluoro-6-hydroxy-4-(trifluoromethyl)benzonitrile

Lieferant: Apollo Scientific
Achtung

PC500303-1G PC500303-250MG
APOSPC500303-1GEA 219 EUR
APOSPC500303-1G APOSPC500303-250MG
2-Fluoro-6-hydroxy-4-(trifluoromethyl)benzonitrile
2-Fluoro-6-hydroxy-4-(trifluoromethyl)benzonitrile
Formel: C₈H₃F₄NO
Molecular Weight: 205,109 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27975490
CAS-Nummer: 1823891-92-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR