2,6-Difluoro-4-(trifluoromethyl)benzonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC500312-1G
PC500312-250MG
APOSPC500312-1GEA
551
EUR
APOSPC500312-1G
APOSPC500312-250MG
2,6-Difluoro-4-(trifluoromethyl)benzonitrile 95%
2,6-Difluoro-4-(trifluoromethyl)benzonitrile
Formel:
C₈H₂F₅N Molecular Weight: 207,1 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27975487 CAS-Nummer: 1803828-56-9 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...