2-Fluoro-5-iodo-4-methylphenol

Lieferant: Apollo Scientific
Achtung

PC500313-1G PC500313-5G
APOSPC500313-1GEA 270 EUR
APOSPC500313-1G APOSPC500313-5G
2-Fluoro-5-iodo-4-methylphenol
2-Fluoro-5-iodo-4-methylphenol
Formel: C₇H₆FIO
Molecular Weight: 252,025 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27975459
CAS-Nummer: 1823935-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR