5-Amino-2-fluoro-4-methylanisole

Lieferant: Apollo Scientific
Achtung

PC500317-1G PC500317-250MG
APOSPC500317-1GEA 511 EUR
APOSPC500317-1G APOSPC500317-250MG
5-Amino-2-fluoro-4-methylanisole
5-Amino-2-fluoro-4-methylanisole
Formel: C₈H₁₀FNO
Molecular Weight: 155,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20714261
CAS-Nummer: 77153-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR