3-Fluoro-2-iodophenol

Lieferant: Apollo Scientific
Achtung

PC500356-1G PC500356-250MG PC500356-5G
APOSPC500356-1GEA 306 EUR
APOSPC500356-1G APOSPC500356-250MG APOSPC500356-5G
3-Fluoro-2-iodophenol
3-Fluoro-2-iodophenol
Formel: C₆H₄FIO
Molecular Weight: 237,998 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08166555
CAS-Nummer: 863870-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR