2-Bromo-5-fluoro-3-methylaniline 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-2-bromo-5-fluorotoluene
PC500365-1G
PC500365-250MG
PC500365-5G
APOSPC500365-1GEA
377
EUR
APOSPC500365-1G
APOSPC500365-250MG
APOSPC500365-5G
2-Bromo-5-fluoro-3-methylaniline 97%
2-Bromo-5-fluoro-3-methylaniline
Formel:
C₇H₇BrFN Molecular Weight: 204.04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27975494 CAS-Nummer: 1463053-93-1 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...