2-Bromo-6-methoxybenzotrifluoride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-3-methoxy-2-(trifluoromethyl)benzene, 3-Bromo-2-(trifluoromethyl)phenyl methyl ether

PC500370-1G PC500370-250MG PC500370-5G
APOSPC500370-1GEA 58 EUR
APOSPC500370-1G APOSPC500370-250MG APOSPC500370-5G
2-Bromo-6-methoxybenzotrifluoride 95%
2-Bromo-6-methoxybenzotrifluoride
Formel: C₈H₆BrF₃O
Molecular Weight: 255.032 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194484
CAS-Nummer: 1214345-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR