3-Chloro-5-(trifluoromethoxy)phenacyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-[3-chloro-5-(trifluoromethoxy)phenyl]ethanone

PC500373-1G PC500373-250MG
APOSPC500373-1GEA 0 EUR
APOSPC500373-1G APOSPC500373-250MG
3-Chloro-5-(trifluoromethoxy)phenacyl bromide
3-Chloro-5-(trifluoromethoxy)phenacyl bromide
Formel: C₉H₅BrClF₃O₂
Molecular Weight: 317.487 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27975441
CAS-Nummer: 1796558-59-2
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR