2-Fluoro-4-hydroxy-5-nitrobenzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

PC500450-1G PC500450-250MG PC500450-5G
APOSPC500450-1GEA 174 EUR
APOSPC500450-1G APOSPC500450-250MG APOSPC500450-5G
2-Fluoro-4-hydroxy-5-nitrobenzaldehyde 95%
2-Fluoro-4-hydroxy-5-nitrobenzaldehyde
Formel: C₇H₄FNO₄
Molecular Weight: 185.109 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27995610
CAS-Nummer: 394-34-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR