2,3-Difluoro-4-iodobenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC500469-1G
PC500469-250MG
PC500469-5G
APOSPC500469-1GEA
75.4
EUR
APOSPC500469-1G
APOSPC500469-250MG
APOSPC500469-5G
2,3-Difluoro-4-iodobenzoic acid
2,3-Difluoro-4-iodobenzoic acid
Formel:
C₇H₃F₂IO₂ Molecular Weight: 283,999 g/mol Schmelzpunkt: 195…197 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368123 CAS-Nummer: 501433-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...