2-(Difluoromethoxy)phenylacetylene

Lieferant: Apollo Scientific
Gefahr

PC500482-1G PC500482-250MG PC500482-5G
APOSPC500482-1GEA 65.4 EUR
APOSPC500482-1G APOSPC500482-250MG APOSPC500482-5G
2-(Difluoromethoxy)phenylacetylene
2-(Difluoromethoxy)phenylacetylene
Formel: C₉H₆F₂O
Molecular Weight: 168,14 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17281928
CAS-Nummer: 1342158-87-5
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR