2-Fluoro-5-nitrobenzenesulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
PC500491-1G
PC500491-250MG
PC500491-5G
APOSPC500491-1GEA
200
EUR
APOSPC500491-1G
APOSPC500491-250MG
APOSPC500491-5G
2-Fluoro-5-nitrobenzenesulphonyl chloride
2-Fluoro-5-nitrobenzenesulphonyl chloride
Formel:
C₆H₃ClFNO₄S Molecular Weight: 239,609 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08443561 CAS-Nummer: 713-21-3 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...