5-Bromo-2-fluorostyrene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-1-fluoro-2-vinylbenze, 4-Bromo-2-ethenyl-1-fluorobenzene
PC500496-1G
PC500496-250MG
PC500496-5G
APOSPC500496-1GEA
58
EUR
APOSPC500496-1G
APOSPC500496-250MG
APOSPC500496-5G
5-Bromo-2-fluorostyrene
5-Bromo-2-fluorostyrene
Formel:
C₈H₆BrF Molecular Weight: 201.036 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD20384566 CAS-Nummer: 221030-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...