3-Bromo-6-chloro-2-fluorobenzyl alcohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Bromo-6-chloro-2-fluorophenyl)methanol
PC500497-1G
PC500497-5G
PC500497-10G
APOSPC500497-1GEA
212
EUR
APOSPC500497-1G
APOSPC500497-5G
APOSPC500497-10G
3-Bromo-6-chloro-2-fluorobenzyl alcohol 98%
3-Bromo-6-chloro-2-fluorobenzyl alcohol
Formel:
C₇H₅BrClFO Molecular Weight: 239,469 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18447606 CAS-Nummer: 886615-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...