4-(Bromomethyl)-2-fluorobenzaldehyde 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Fluoro-4-formylbenzyl bromide
PC500500-1G
PC500500-250MG
PC500500-5G
APOSPC500500-1GEA
249
EUR
APOSPC500500-1G
APOSPC500500-250MG
APOSPC500500-5G
4-(Bromomethyl)-2-fluorobenzaldehyde 95%
4-(Bromomethyl)-2-fluorobenzaldehyde
Formel:
C₈H₆BrFO Molecular Weight: 217,035 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18378725 CAS-Nummer: 1379307-40-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...