4-Bromo-3-(trifluoromethyl)phenacyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromo-5-(bromoacetyl)benzotrifluoride, 2-Bromo-1-[4-bromo-3-(trifluoromethyl)phenyl]ethan-1-one
PC500506-1G
PC500506-250MG
PC500506-5G
APOSPC500506-1GEA
406
EUR
APOSPC500506-1G
APOSPC500506-250MG
APOSPC500506-5G
4-Bromo-3-(trifluoromethyl)phenacyl bromide
4-Bromo-3-(trifluoromethyl)phenacyl bromide
Formel:
C₉H₅Br₂F₃O Molecular Weight: 345,939 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22491116 CAS-Nummer: 1523196-85-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...