3'-Bromo-6'-chloro-2'-fluoroacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Bromo-6-chloro-2-fluorophenyl)ethanone

PC500508-1G PC500508-250MG
APOSPC500508-1GEA 306 EUR
APOSPC500508-1G APOSPC500508-250MG
3'-Bromo-6'-chloro-2'-fluoroacetophenone
3'-Bromo-6'-chloro-2'-fluoroacetophenone
Formel: C₈H₅BrClFO
Lagertemperatur: Raumtemperatur
MDL: MFCD28040519
CAS-Nummer: 1541643-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR