4-Bromo-2-(trifluoromethyl)benzyl alcohol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-2-(trifluoromethyl)benzenemethanol
PC500516-1G
PC500516-25G
PC500516-5G
PC500516-10G
APOSPC500516-1GEA
58
EUR
APOSPC500516-1G
APOSPC500516-25G
APOSPC500516-5G
APOSPC500516-10G
4-Bromo-2-(trifluoromethyl)benzyl alcohol 95%
4-Bromo-2-(trifluoromethyl)benzyl alcohol
Formel:
C₈H₆BrF₃O Molecular Weight: 255.032 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12761598 CAS-Nummer: 932390-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...