3-(Bromomethyl)-4-fluorobenzaldehyde

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Fluoro-5-formylbenzyl bromide

PC500522-1G PC500522-250MG
APOSPC500522-1GEA 348 EUR
APOSPC500522-1G APOSPC500522-250MG
3-(Bromomethyl)-4-fluorobenzaldehyde
3-(Bromomethyl)-4-fluorobenzaldehyde
Formel: C₈H₆BrFO
Molecular Weight: 217,035 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18378686
CAS-Nummer: 1379371-56-8
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR