2-(Bromomethyl)-3-fluorobenzonitrile
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
α-Bromo-3-fluoro-o-tolunitrile, 2-Cyano-6-fluorobenzyl bromide
PC500524-1G
PC500524-250MG
PC500524-5G
APOSPC500524-1GEA
66.7
EUR
APOSPC500524-1G
APOSPC500524-250MG
APOSPC500524-5G
2-(Bromomethyl)-3-fluorobenzonitrile
2-(Bromomethyl)-3-fluorobenzonitrile
Formel:
C₈H₅BrFN Molecular Weight: 214,034 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12025416 CAS-Nummer: 635723-84-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...