2'-Amino-6'-methoxy-2,2,2-trifluoroacetophenone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Amino-6-methoxyphenyl)-2,2,2-trifluoroethanone, 3-Methoxy-2-(trifluoroacetyl)aniline, 1-(2-Amino-6-methoxyphenyl)-2,2,2-trifluoroethan-1-one, 3-Amino-2-(trifluoroacetyl)anisole
PC50053-1G
PC50053-250MG
APOSPC50053-1GEA
1110
EUR
APOSPC50053-1G
APOSPC50053-250MG
2'-Amino-6'-methoxy-2,2,2-trifluoroacetophenone
2'-Amino-6'-methoxy-2,2,2-trifluoroacetophenone
Formel:
C₉H₈F₃NO₂ Molecular Weight: 219,161 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18071075 CAS-Nummer: 1233967-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...