2-(Bromomethyl)-3-fluorobenzaldehyde 95%

Lieferant: Apollo Scientific
Gefahr

PC500534-1G PC500534-250MG PC500534-5G
APOSPC500534-1GEA 183 EUR
APOSPC500534-1G APOSPC500534-250MG APOSPC500534-5G
2-(Bromomethyl)-3-fluorobenzaldehyde 95%
2-(Bromomethyl)-3-fluorobenzaldehyde
Formel: C₈H₆BrFO
Molecular Weight: 217.035 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18205512
CAS-Nummer: 1379312-99-8
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR