3-(Bromomethyl)-5-fluorobenzaldehyde
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Fluoro-5-formylbenzyl bromide
PC500537-1G
PC500537-250MG
APOSPC500537-1GEA
493
EUR
APOSPC500537-1G
APOSPC500537-250MG
3-(Bromomethyl)-5-fluorobenzaldehyde
3-(Bromomethyl)-5-fluorobenzaldehyde
Formel:
C₈H₆BrFO Molecular Weight: 217,035 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18378635 CAS-Nummer: 1379358-86-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...