2-(Bromomethyl)-5-fluorobenzaldehyde
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Fluoro-2-formylbenzyl bromide, α-Bromo-5-fluoro-o-tolualdehyde
PC500562-1G
PC500562-250MG
PC500562-5G
APOSPC500562-1GEA
905
EUR
APOSPC500562-1G
APOSPC500562-250MG
APOSPC500562-5G
2-(Bromomethyl)-5-fluorobenzaldehyde
2-(Bromomethyl)-5-fluorobenzaldehyde
Formel:
C₈H₆BrFO Molecular Weight: 217.035 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18205514 CAS-Nummer: 945995-09-5 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...