5-(Bromomethyl)-2-fluorobenzaldehyde
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Fluoro-3-formylbenzyl bromide, α-Bromo-6-fluoro-m-tolualdehyde
PC500573-1G
PC500573-250MG
PC500573-5G
APOSPC500573-1GEA
435
EUR
APOSPC500573-1G
APOSPC500573-250MG
APOSPC500573-5G
5-(Bromomethyl)-2-fluorobenzaldehyde
5-(Bromomethyl)-2-fluorobenzaldehyde
Formel:
C₈H₆BrFO Molecular Weight: 217,035 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD28096626 CAS-Nummer: 1820717-85-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...