3-Bromo-2-fluoro-6-hydroxy-5-nitrobenzotrifluoride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-6-nitro-α,α,α,3-tetrafluoro-o-cresol, 4-Bromo-3-fluoro-6-nitro-2-(trifluoromethyl)phenol

PC50066-1G PC50066-250MG
APOSPC50066-1GEA 372 EUR
APOSPC50066-1G APOSPC50066-250MG
3-Bromo-2-fluoro-6-hydroxy-5-nitrobenzotrifluoride 97%
3-Bromo-2-fluoro-6-hydroxy-5-nitrobenzotrifluoride
Formel: C₇H₂BrF₄NO₃
Lagertemperatur: Kühlschrank
MDL: MFCD25541892
CAS-Nummer: 1445995-81-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR