2-Amino-6-bromo-3-nitrobenzotrifluoride 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-6-nitro-α,α,α-trifluoro-o-toluidine, 3-Bromo-6-nitro-2-(trifluoromethyl)aniline

PC50067-1G PC50067-250MG
APOSPC50067-1GEA 522 EUR
APOSPC50067-1G APOSPC50067-250MG
2-Amino-6-bromo-3-nitrobenzotrifluoride 96%
2-Amino-6-bromo-3-nitrobenzotrifluoride
Formel: C₇H₄BrF₃N₂O₂
Molecular Weight: 285,018 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD25541893
CAS-Nummer: 1445995-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR