Methyl 4-cyano-2-fluorobenzoate

Lieferant: Apollo Scientific
Achtung

PC500686-25G PC500686-5G PC500686-50G
APOSPC500686-25GEA 345 EUR
APOSPC500686-25G APOSPC500686-5G APOSPC500686-50G
Methyl 4-cyano-2-fluorobenzoate
Methyl 4-cyano-2-fluorobenzoate
Formel: C₉H₆FNO₂
Molecular Weight: 179.148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04335187
CAS-Nummer: 175596-01-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR